missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzyl Chloride 98.0+%, TCI America™
$180.97 - $180.97
Chemical Identifiers
| CAS | 2014-83-7 |
|---|---|
| Molecular Formula | C7H5Cl3 |
| Molecular Weight (g/mol) | 195.467 |
| MDL Number | MFCD00000897 |
| InChI Key | LBOBESSDSGODDD-UHFFFAOYSA-N |
| Synonym | 2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene |
| PubChem CID | 74832 |
| IUPAC Name | 1,3-dichloro-2-(chloromethyl)benzene |
| SMILES | C1=CC(=C(C(=C1)Cl)CCl)Cl |
Chemical Identifiers
| 2014-83-7 | |
| 195.467 | |
| LBOBESSDSGODDD-UHFFFAOYSA-N | |
| 74832 | |
| C1=CC(=C(C(=C1)Cl)CCl)Cl |
| C7H5Cl3 | |
| MFCD00000897 | |
| 2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene | |
| 1,3-dichloro-2-(chloromethyl)benzene |
Specifications
| 2014-83-7 | |
| White-Yellow | |
| C7H5Cl3 | |
| 25 g | |
| 2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene | |
| C1=CC(=C(C(=C1)Cl)CCl)Cl | |
| 195.467 | |
| 195.47 | |
| Crystalline Lumps |
| 38°C | |
| 119°C | |
| MFCD00000897 | |
| 3261 | |
| LBOBESSDSGODDD-UHFFFAOYSA-N | |
| 1,3-dichloro-2-(chloromethyl)benzene | |
| 74832 | |
| ≥98.0% (GC) | |
| 2,6-Dichlorobenzyl Chloride |
Safety and Handling
TSCA : No