missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America D206425G
Specifications
| 2,6-Dichlorobenzoyl Chloride | |
| Yellow | |
| C7H3Cl3O | |
| 25 g | |
| benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043 | |
| C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl | |
| 209.45 | |
| 209.45 | |
| Liquid |
| 4659-45-4 | |
| 127°C | |
| MFCD00000662 | |
| 3265 | |
| JBLIDPPHFGWTKU-UHFFFAOYSA-N | |
| 2,6-dichlorobenzoyl chloride | |
| 78392 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4659-45-4 | |
| 209.45 | |
| JBLIDPPHFGWTKU-UHFFFAOYSA-N | |
| 78392 | |
| C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl |
| C7H3Cl3O | |
| MFCD00000662 | |
| benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043 | |
| 2,6-dichlorobenzoyl chloride |
Safety and Handling
EINECSNumber : (3)-0062&(3)-1409
TSCA : No