missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D2064500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2064500G 500 g
D206425G 25 g
D2064100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2064500G Supplier TCI America Supplier No. D2064500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4659-45-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000662
InChI Key JBLIDPPHFGWTKU-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
PubChem CID 78392
IUPAC Name 2,6-dichlorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 127°C
Quantity 500 g
UN Number 3265
Formula Weight 209.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dichlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA No