missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoyl chloride, 98%

Catalog No. AAA1483918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1483918 50 g
AAA1483909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1483918 Supplier Thermo Scientific Chemicals Supplier No. A1483918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4659-45-4 | C7H3Cl3O | 209.45 g/mol

2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

Solubility
Miscible with alcohol, ether and acetone. Slightly miscible with heptane. Immiscible with water.

Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents and strong bases.

Chemical Identifiers

CAS 4659-45-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000662
InChI Key JBLIDPPHFGWTKU-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
PubChem CID 78392
IUPAC Name 2,6-dichlorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl

Specifications

Melting Point 15°C to 18°C
Density 1.463
Boiling Point 130°C to 132°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 50 g
UN Number UN3265
Beilstein 639531
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol,ether and acetone. Slightly miscible with heptane. Immiscible with water.
Formula Weight 209.46
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 225-102-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only