missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzonitrile 99.0+%, TCI America™
$61.51 - $61.51
Chemical Identifiers
| CAS | 1194-65-6 |
|---|---|
| Molecular Formula | C7H3Cl2N |
| Molecular Weight (g/mol) | 172.008 |
| MDL Number | MFCD00001781 |
| InChI Key | YOYAIZYFCNQIRF-UHFFFAOYSA-N |
| Synonym | dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 |
| PubChem CID | 3031 |
| ChEBI | CHEBI:943 |
| IUPAC Name | 2,6-dichlorobenzonitrile |
| SMILES | C1=CC(=C(C(=C1)Cl)C#N)Cl |
Chemical Identifiers
| 1194-65-6 | |
| 172.008 | |
| YOYAIZYFCNQIRF-UHFFFAOYSA-N | |
| 3031 | |
| 2,6-dichlorobenzonitrile |
| C7H3Cl2N | |
| MFCD00001781 | |
| dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 | |
| CHEBI:943 | |
| C1=CC(=C(C(=C1)Cl)C#N)Cl |
Specifications
| 1194-65-6 | |
| White | |
| MFCD00001781 | |
| 3077 | |
| YOYAIZYFCNQIRF-UHFFFAOYSA-N | |
| 2,6-dichlorobenzonitrile | |
| 3031 | |
| 172.01 | |
| Crystalline Powder |
| 145°C | |
| C7H3Cl2N | |
| 25 g | |
| dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 | |
| C1=CC(=C(C(=C1)Cl)C#N)Cl | |
| 172.008 | |
| CHEBI:943 | |
| ≥99.0% (GC) | |
| 2,6-Dichlorobenzonitrile |
Safety and Handling
EINECSNumber : (3)-4103
RTECSNumber : DI3500000
TSCA : No