accessibility menu, dialog, popup

UserName

ClClN

CAS RN 1194-65-6

IUPAC Name: 2,6-dichlorobenzonitrile
Synonyms: prefix D caswell no. 297 DU-sprex surfassol dichlobanil cyclomec DBN dyclomec dichlobenil niagara 5,996
Molecular Weight (g/mol): 172.01
Molecular Formula: C7H3Cl2N
InChi Key: YOYAIZYFCNQIRF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1C#N
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)