missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzonitrile, 98%

Catalog No. AAA1498622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1498622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1498622 Supplier Thermo Scientific Chemicals Supplier No. A1498622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1194-65-6 | C7H3Cl2N | 172.008 g/mol

2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

Solubility
Soluble in water (0.018g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1194-65-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00001781
InChI Key YOYAIZYFCNQIRF-UHFFFAOYSA-N
Synonym dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133
PubChem CID 3031
ChEBI CHEBI:943
IUPAC Name 2,6-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)Cl

Specifications

Melting Point 143°C to 147°C
Boiling Point 270°C
Odor Aromatic
Quantity 100 g
UN Number UN3077
Beilstein 1909167
Merck Index 14,3042
Solubility Information Soluble in water (0.018g/L).
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzonitrile
Show More Show Less
Hazard Category H312
Hazard Statement GHS H Statement
H312
Harmful in contact with skin.
Precautionary Statement P280-P302+P352-P312-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-787-5
RTECSNumber DI3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only