missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoic acid, 97%

Catalog No. p-7080826
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1054214 25 g
AAA1054222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1054214 Supplier Thermo Scientific Chemicals Supplier No. A1054214
Only null left

CAS: 50-30-6 | C7H4Cl2O2 | 191.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-30-6
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002418
InChI Key MRUDNSFOFOQZDA-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid
PubChem CID 5758
ChEBI CHEBI:48623
IUPAC Name 2,6-dichlorobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 141°C to 145°C
Quantity 25 g
Beilstein 973858
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-025-9
RTECSNumber DG7000000
TSCA Yes

RUO – Research Use Only