missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzenethiol 95.0+%, TCI America™

Catalog No. p-7127003
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19045G 5 g
D190425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19045G Supplier TCI America Supplier No. D19045G

Chemical Identifiers

CAS 24966-39-0
Molecular Formula C6H3Cl2S
Molecular Weight (g/mol) 178.05
MDL Number MFCD00004833
InChI Key JBISHCXLCGVPGW-UHFFFAOYSA-M
Synonym 2,6-dichlorothiophenol, 2,6-dichlorobenzene-1-thiol, 2,6-dichloro thiophenol, benzenethiol, 2,6-dichloro, pubchem6802, 2,6-dichloro-benzenethiol, acmc-209gg5, benzenethiol,2,6-dichloro
PubChem CID 90671
IUPAC Name (2,6-dichlorophenyl)sulfanide
SMILES [S-]C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzenethiol