missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzamide, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 2008-58-4 |
|---|---|
| Molecular Formula | C7H5Cl2NO |
| Molecular Weight (g/mol) | 190.03 |
| MDL Number | MFCD00007975 |
| InChI Key | JHSPCUHPSIUQRB-UHFFFAOYSA-N |
| Synonym | benzamide, 2,6-dichloro, 2,6-bam, unii-e9jwf529eb, 2,6-dichlorobenzoic acid amide, bam, e9jwf529eb, benzamide,6-dichloro, dsstox_cid_2170, acmc-209f5j, dsstox_rid_76511 |
| PubChem CID | 16183 |
| ChEBI | CHEBI:28435 |
| IUPAC Name | 2,6-dichlorobenzamide |
| SMILES | C1=CC(=C(C(=C1)Cl)C(=O)N)Cl |
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC161690250
|
N/A | 25g | Glass bottle |
N/A
|
|
||||
|
AC161691000
|
N/A | 100g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C7H5Cl2NO | |
| MFCD00007975 | |
| benzamide, 2,6-dichloro, 2,6-bam, unii-e9jwf529eb, 2,6-dichlorobenzoic acid amide, bam, e9jwf529eb, benzamide,6-dichloro, dsstox_cid_2170, acmc-209f5j, dsstox_rid_76511 | |
| CHEBI:28435 | |
| C1=CC(=C(C(=C1)Cl)C(=O)N)Cl |
Specifications
| 2008-58-4 | |
| 100.0 | |
| 96% min. (GC) | |
| Cl2C6H3CONH2 | |
| 09, I, 141 | |
| JHSPCUHPSIUQRB-UHFFFAOYSA-N | |
| 2,6-dichlorobenzamide | |
| 16183 | |
| 190.03 | |
| 2, 6-Dichlorobenzamide, 97% |
| 96.0 | |
| Authentic | |
| C7H5Cl2NO | |
| MFCD00007975 | |
| benzamide, 2,6-dichloro, 2,6-bam, unii-e9jwf529eb, 2,6-dichlorobenzoic acid amide, bam, e9jwf529eb, benzamide,6-dichloro, dsstox_cid_2170, acmc-209f5j, dsstox_rid_76511 | |
| C1=CC(=C(C(=C1)Cl)C(=O)N)Cl | |
| 190.03 | |
| CHEBI:28435 | |
| 97% |
Safety and Handling
EINECSNumber : 217-918-4
TSCA : TSCA
RUO – Research Use Only