missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetophenone 98.0+%, TCI America™

Catalog No. p-7128107
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41191G 1 g
D41195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41191G Supplier TCI America Supplier No. D41191G
Only null left

Chemical Identifiers

CAS 2040-05-3
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00016339
InChI Key HYBDSXBLGCQKRE-UHFFFAOYSA-N
Synonym 2',6'-dichloroacetophenone, 1-2,6-dichlorophenyl ethanone, 2,6-dichloroacetophenone, 1-2,6-dichlorophenyl ethan-1-one, ethanone, 1-2,6-dichlorophenyl, 2,6-dichloro acetophenone, 1-acetyl-2,6-dichlorobenzene, zlchem 1084, pubchem12362, acmc-1cobf
PubChem CID 74877
IUPAC Name 1-(2,6-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1Cl)Cl

Specifications

Melting Point 44°C
Boiling Point 141°C
Quantity 1 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2′,6′-Dichloroacetophenone