Learn More
2',6'-Dichloroacetophenone, 98%
CAS: 2040-05-3 | C8H6Cl2O | 189.04 g/mol
Supplier: Thermo Scientific Chemicals L0634803
Description
2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
| 2',6'-Dichloroacetophenone | |
| 41°C to 44°C | |
| C11H14O | |
| 1 g | |
| 2',4',5'-trimethylacetophenone, 1-2,4,5-trimethylphenyl ethanone, 1-2,4,5-trimethylphenyl ethan-1-one, acetophenone, 2',4',5'-trimethyl, ethanone, 1-2,4,5-trimethylphenyl, 1-acetyl-2,4,5-trimethylbenzene, acmc-20aofe, 2',5'-trimethylacetophenone, 2,4,5-trimethyl-acetophenone, acetophenone,4',5'-trimethyl | |
| GENBEGZNCBFHSU-UHFFFAOYSA-N | |
| 1-(2,4,5-trimethylphenyl)ethanone | |
| 74878 | |
| 98% |
| 2040-05-3 | |
| 176°C to 180°C (83 mmHg) | |
| MFCD00016339 | |
| 2440104 | |
| Insoluble in water. | |
| CC1=CC(=C(C=C1C(=O)C)C)C | |
| 162.23 | |
| 189.04 |
Chemical Identifiers
| 2040-05-3 | |
| MFCD00016339 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 218-035-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only