missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-7-deazapurine 98.0+%, TCI America™

Catalog No. p-7128198
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D42841G 1 g
D42845G 5 g
2 options

Catalog No. D42841G

Supplier: TCI America D42841G

Only null left

Chemical Identifiers

CAS 90213-66-4
Molecular Formula C6H3Cl2N3
Molecular Weight (g/mol) 188.011
MDL Number MFCD08059278
InChI Key GHXBPCSSQOKKGB-UHFFFAOYSA-N
Synonym 2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
PubChem CID 14116871
IUPAC Name 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=CNC2=C1C(=NC(=N2)Cl)Cl

Specifications

Melting Point 251°C
Color Yellow
Quantity 1 g
Formula Weight 188.01
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-7-deazapurine