missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-5-fluoronicotinic Acid 97.0+%, TCI America™

Catalog No. p-7127401
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26105G 5 g
D261025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26105G Supplier TCI America Supplier No. D26105G
Only null left

Chemical Identifiers

CAS 82671-06-5
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.985
MDL Number MFCD00799517
InChI Key LTDGKGCHRNNCAC-UHFFFAOYSA-N
Synonym 2,6-dichloro-5-fluoronicotinic acid, 2,6-dichloro-5-fluoro-nicotinic acid, 2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid, 2,6-dichloro-5-fluoro nicotinic acid, 3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro, pubchem10943, acmc-1bluq, ksc448a5j, 2,6-dichloro-5-fluoro-nicotinicacid
PubChem CID 2733659
IUPAC Name 2,6-dichloro-5-fluoropyridine-3-carboxylic acid
SMILES C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 209.99
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-5-fluoronicotinic Acid