missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-(trifluoromethyl)pyridine 97.0+%, TCI America™
$73.16 - $246.43
Chemical Identifiers
| CAS | 39890-98-7 |
|---|---|
| Molecular Formula | C6H2Cl2F3N |
| Molecular Weight (g/mol) | 215.98 |
| MDL Number | MFCD00042246 |
| InChI Key | KVNQWVYYVLCZKK-UHFFFAOYSA-N |
| Synonym | 2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-4-trifluoromethyl-pyridine, 4-trifluoromethyl-2,6-dichloropyridine, 2,6-dichloro-4-trifluoromethylpyridine, acmc-1air6, abbypharma ap-30-1899, pyridine, 2,6-dichloro-4-trifluoromethyl |
| PubChem CID | 520968 |
| IUPAC Name | 2,6-dichloro-4-(trifluoromethyl)pyridine |
| SMILES | FC(F)(F)C1=CC(Cl)=NC(Cl)=C1 |
Chemical Identifiers
| 39890-98-7 | |
| 215.98 | |
| KVNQWVYYVLCZKK-UHFFFAOYSA-N | |
| 520968 | |
| FC(F)(F)C1=CC(Cl)=NC(Cl)=C1 |
| C6H2Cl2F3N | |
| MFCD00042246 | |
| 2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-4-trifluoromethyl-pyridine, 4-trifluoromethyl-2,6-dichloropyridine, 2,6-dichloro-4-trifluoromethylpyridine, acmc-1air6, abbypharma ap-30-1899, pyridine, 2,6-dichloro-4-trifluoromethyl | |
| 2,6-dichloro-4-(trifluoromethyl)pyridine |
Specifications
| 39890-98-7 | |
| 57°C | |
| MFCD00042246 | |
| 2810 | |
| KVNQWVYYVLCZKK-UHFFFAOYSA-N | |
| 2,6-dichloro-4-(trifluoromethyl)pyridine | |
| 520968 | |
| ≥97.0% (GC) | |
| 2,6-Dichloro-4-(trifluoromethyl)pyridine |
| Yellow | |
| C6H2Cl2F3N | |
| 1 g | |
| 2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-4-trifluoromethyl-pyridine, 4-trifluoromethyl-2,6-dichloropyridine, 2,6-dichloro-4-trifluoromethylpyridine, acmc-1air6, abbypharma ap-30-1899, pyridine, 2,6-dichloro-4-trifluoromethyl | |
| FC(F)(F)C1=CC(Cl)=NC(Cl)=C1 | |
| 215.98 | |
| 215.98 | |
| Liquid |
Safety and Handling
TSCA : No