missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. D38025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38025G 5 g
D380225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38025G Supplier TCI America Supplier No. D38025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99479-66-0
Molecular Formula C7H4Cl2F3NO
Molecular Weight (g/mol) 246.01
MDL Number MFCD00190128
InChI Key FKISQWQHZULEEG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
PubChem CID 688247
IUPAC Name 2,6-dichloro-4-(trifluoromethoxy)aniline
SMILES NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl

Specifications

Melting Point 33°C
Quantity 5 g
Formula Weight 246.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)aniline