missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-pyridinemethanol 98.0+%, TCI America™

Catalog No. D44235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44235G 5 g
D44231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44235G Supplier TCI America Supplier No. D44235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101990-69-6 | C6H5Cl2NO | 178.012 g/mol

Chemical Identifiers

CAS 101990-69-6
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD00052638
InChI Key YDGJTFCKLFLWFM-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine-4-methanol, 2,6-dichloropyridin-4-yl methanol, 2,6-dichloro-4-hydroxymethyl pyridine, 2,6-dichloro-4-pyridyl methanol, 4-pyridinemethanol, 2,6-dichloro, 4-pyridinemethanol,2,6-dichloro, 2,6-dichloro-4-pyridinyl methanol, 2,6-dichloro-pyridin-4-yl-methanol, 2,6-dichloro-4-pyridinemethanol, acmc-209zae
PubChem CID 2794840
IUPAC Name (2,6-dichloropyridin-4-yl)methanol
SMILES C1=C(C=C(N=C1Cl)Cl)CO

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 178.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-pyridinemethanol