missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-nitroaniline 97.0+%, TCI America™
$67.11 - $656.14
Chemical Identifiers
| CAS | 99-30-9 |
|---|---|
| Molecular Formula | C6H4Cl2N2O2 |
| Molecular Weight (g/mol) | 207.01 |
| MDL Number | MFCD00007677 |
| InChI Key | BIXZHMJUSMUDOQ-UHFFFAOYSA-N |
| Synonym | dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide |
| PubChem CID | 7430 |
| ChEBI | CHEBI:27864 |
| IUPAC Name | 2,6-dichloro-4-nitroaniline |
| SMILES | C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-] |
Chemical Identifiers
| 99-30-9 | |
| 207.01 | |
| BIXZHMJUSMUDOQ-UHFFFAOYSA-N | |
| 7430 | |
| 2,6-dichloro-4-nitroaniline |
| C6H4Cl2N2O2 | |
| MFCD00007677 | |
| dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide | |
| CHEBI:27864 | |
| C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-] |
Specifications
| 99-30-9 | |
| 130°C | |
| MFCD00007677 | |
| dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide | |
| C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-] | |
| 207.01 | |
| CHEBI:27864 | |
| ≥97.0% (GC) | |
| 2,6-Dichloro-4-nitroaniline |
| 193°C | |
| C6H4Cl2N2O2 | |
| 25 g | |
| BIXZHMJUSMUDOQ-UHFFFAOYSA-N | |
| 2,6-dichloro-4-nitroaniline | |
| 7430 | |
| 207.01 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0423
RTECSNumber : BX2975000
TSCA : Yes