accessibility menu, dialog, popup

UserName

CAS RN 99-30-9

IUPAC Name: 2,6-dichloro-4-nitroaniline
Synonyms: batran
Molecular Weight (g/mol): 207.01
Molecular Formula: C6H4Cl2N2O2
InChi Key: BIXZHMJUSMUDOQ-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)