missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroaniline 97.0+%, TCI America™

Catalog No. D038525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038525G 25 g
D0385500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D038525G Supplier TCI America Supplier No. D038525G

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007677
InChI Key BIXZHMJUSMUDOQ-UHFFFAOYSA-N
Synonym dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide
PubChem CID 7430
ChEBI CHEBI:27864
IUPAC Name 2,6-dichloro-4-nitroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]

Specifications

Melting Point 193°C
Boiling Point 130°C
Quantity 25 g
Formula Weight 207.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-nitroaniline
EINECSNumber (3)-0423
RTECSNumber BX2975000
TSCA Yes