missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroaniline, 95%

Catalog No. AAB2428336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2428336 500 g
AAB2428314 25 g
AAB2428322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2428336 Supplier Thermo Scientific Chemicals Supplier No. B2428336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 99-30-9 | C6H4Cl2N2O2 | 207.01 g/mol

It is used as dyestuff, agricultural and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007677
InChI Key BIXZHMJUSMUDOQ-UHFFFAOYSA-N
Synonym dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide
PubChem CID 7430
ChEBI CHEBI:27864
IUPAC Name 2,6-dichloro-4-nitroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]

Specifications

Melting Point 185°C to 192°C
Boiling Point 130°C (2 mmHg)
Odor Recognizable
Quantity 500 g
Beilstein 1459581
Solubility Information Insoluble in water.
Formula Weight 207.02
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-nitroaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-746-4
RTECSNumber BX2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only