missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-iodopyridine, 97%

CAS: 98027-84-0 | C5H2Cl2IN | 273.882 g/mol

Supplier:  Thermo Scientific Chemicals H2694606

 View more versions of this product

Catalog No. AAH2694606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Dichloro-4-iodopyridine
161°C to 165°C
MFCD07368400
Light sensitive
NGSKFMPSBUAUNE-UHFFFAOYSA-N
2,6-dichloro-4-iodopyridine
11737393
97%
98027-84-0
C5H2Cl2IN
5 g
2,6-dichloro-4-iodo-pyridine, 2 6-dichloro-4-iodopyridine, pyridine, 2,6-dichloro-4-iodo, pubchem17648, 2,6 dichloro-4-iodo pyridine, 2,6-dichloro-4-iodopyridine,, 4-iodo-2,6-dichloropyridine, 2,6-dichloro-4-iodopyridine
C1=C(C=C(N=C1Cl)Cl)I
273.882
273.89

Chemical Identifiers

98027-84-0
273.882
NGSKFMPSBUAUNE-UHFFFAOYSA-N
11737393
C1=C(C=C(N=C1Cl)Cl)I
C5H2Cl2IN
MFCD07368400
2,6-dichloro-4-iodo-pyridine, 2 6-dichloro-4-iodopyridine, pyridine, 2,6-dichloro-4-iodo, pubchem17648, 2,6 dichloro-4-iodo pyridine, 2,6-dichloro-4-iodopyridine,, 4-iodo-2,6-dichloropyridine, 2,6-dichloro-4-iodopyridine
2,6-dichloro-4-iodopyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only