missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-iodopyridine, 97%

Catalog No. p-7029479
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH2694603 1 g
AAH2694606 5 g
2 options

Catalog No. AAH2694603

Supplier: Thermo Scientific Chemicals H2694603

Only null left

CAS: 98027-84-0 | C5H2Cl2IN | 273.882 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98027-84-0
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.882
MDL Number MFCD07368400
InChI Key NGSKFMPSBUAUNE-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-iodo-pyridine, 2 6-dichloro-4-iodopyridine, pyridine, 2,6-dichloro-4-iodo, pubchem17648, 2,6 dichloro-4-iodo pyridine, 2,6-dichloro-4-iodopyridine,, 4-iodo-2,6-dichloropyridine, 2,6-dichloro-4-iodopyridine
PubChem CID 11737393
IUPAC Name 2,6-dichloro-4-iodopyridine
SMILES C1=C(C=C(N=C1Cl)Cl)I

Specifications

Melting Point 161°C to 165°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 273.89
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-4-iodopyridine
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only