missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-fluorophenol, 98%

CAS: 392-71-2 | C6H3Cl2FO | 180.987 g/mol

$78.92 - $1094.49

Chemical Identifiers

CAS 392-71-2
Molecular Formula C6H3Cl2FO
Molecular Weight (g/mol) 180.987
MDL Number MFCD00010675
InChI Key BOJVIFKSTRCIRJ-UHFFFAOYSA-N
PubChem CID 94950
IUPAC Name 2,6-dichloro-4-fluorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1619306
View Documents
Thermo Scientific Chemicals
A1619306
5 g
Each for $78.92
Only null left
 
AAA1619322
View Documents
Thermo Scientific Chemicals
A1619322
100 g
Each for $1,094.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

392-71-2
180.987
BOJVIFKSTRCIRJ-UHFFFAOYSA-N
2,6-dichloro-4-fluorophenol
C6H3Cl2FO
MFCD00010675
94950
C1=C(C=C(C(=C1Cl)O)Cl)F

Specifications

392-71-2
142°C (100 mmHg)
Phenol-like
MFCD00010675
UN3261
phenol, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluoro-phenol, pubchem2871, acmc-1ai43, 4-fluoro-2,6-dichlorophenol, bojvifkstrcirj-uhfffaoysa, phenol,2,6-dichloro-4-fluoro, attercop-chm at126185
C1=C(C=C(C(=C1Cl)O)Cl)F
180.987
180.99
2,6-Dichloro-4-fluorophenol
47°C to 51°C
103°C (217°F)
C6H3Cl2FO
5 g
2046856
BOJVIFKSTRCIRJ-UHFFFAOYSA-N
2,6-dichloro-4-fluorophenol
94950
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only