missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-fluorophenol, 98%

Catalog No. AAA1619322
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1619322 100 g
AAA1619306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1619322 Supplier Thermo Scientific Chemicals Supplier No. A1619322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392-71-2 | C6H3Cl2FO | 180.987 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 392-71-2
Molecular Formula C6H3Cl2FO
Molecular Weight (g/mol) 180.987
MDL Number MFCD00010675
InChI Key BOJVIFKSTRCIRJ-UHFFFAOYSA-N
PubChem CID 94950
IUPAC Name 2,6-dichloro-4-fluorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)F

Specifications

Melting Point 47°C to 51°C
Boiling Point 142°C (100 mmHg)
Flash Point 103°C (217°F)
Odor Phenol-like
Quantity 100 g
UN Number UN3261
Beilstein 2046856
Synonym phenol, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluoro-phenol, pubchem2871, acmc-1ai43, 4-fluoro-2,6-dichlorophenol, bojvifkstrcirj-uhfffaoysa, phenol,2,6-dichloro-4-fluoro, attercop-chm at126185
Formula Weight 180.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-4-fluorophenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only