missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals

Catalog No. AAH32374MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32374MD 250 mg
AAH3237403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32374MD Supplier Thermo Scientific Chemicals Supplier No. H32374MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1099597-81-5
MDL Number MFCD11100517

Specifications

Melting Point 95°C to 99°C
Molecular Formula C11H5Cl2FN2O
Quantity 250 mg
Formula Weight 271.08
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only