accessibility menu, dialog, popup

UserName

ClClFONN

CAS RN 1099597-81-5

IUPAC Name: 2,4-dichloro-6-(4-fluorobenzoyl)pyrimidine
Synonyms: 2,4-dichloro-6-(4-fluorobenzoyl)pyrimidine
Molecular Weight (g/mol): 271.07
Molecular Formula: C11H5Cl2FN2O
InChi Key: QIULZDCQGAXJIB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=NC(Cl)=NC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results