missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAL2000303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2000303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2000303 Supplier Thermo Scientific Chemicals Supplier No. L2000303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55304-75-1 | Molecular Formula: C6H2Cl2F3N | Molecular Weight: 215.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55304-75-1
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042245
InChI Key UPWAAFFFSGQECJ-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-trifluoromethyl pyridine, pyridine, 2,6-dichloro-3-trifluoromethyl, 2,6-dichloro-3-trifluoromethyl-pyridine, 2,6-dichloro-3-trifluoromethyl pyridine, puriss, pubchem9238, acmc-20a2ib, 2,6-dichloro-5-trifluoromethylpyridine
PubChem CID 1810337
IUPAC Name 2,6-dichloro-3-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1Cl

Specifications

Boiling Point 194°C to 196°C
Refractive Index 1.4845
Quantity 1 g
UN Number UN2810
Beilstein 1570287
Formula Weight 215.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 259-585-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only