CAS RN 55304-75-1
IUPAC Name:
2,6-dichloro-3-(trifluoromethyl)pyridine
Synonyms:
2,6-dichloro-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
215.98
Molecular Formula:
C6H2Cl2F3N
InChi Key:
UPWAAFFFSGQECJ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(Cl)N=C1Cl
1
–
1
of
1
results
1
Promotions Available
2,6-Dichloro-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
2,6-Dichloro-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
CAS: 55304-75-1 Molecular Formula: C6H2Cl2F3N Molecular Weight (g/mol): 215.98 MDL Number: MFCD00042245 InChI Key: UPWAAFFFSGQECJ-UHFFFAOYSA-N Synonym: 2,6-dichloro-3-trifluoromethyl pyridine,pyridine, 2,6-dichloro-3-trifluoromethyl,2,6-dichloro-3-trifluoromethyl-pyridine,2,6-dichloro-3-trifluoromethyl pyridine, puriss,pubchem9238,acmc-20a2ib,2,6-dichloro-5-trifluoromethylpyridine PubChem CID: 1810337 IUPAC Name: 2,6-dichloro-3-(trifluoromethyl)pyridine SMILES: FC(F)(F)C1=CC=C(Cl)N=C1Cl