missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitrotoluene, 99%

Catalog No. AAB2052009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2052009 10 g
AAB2052018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2052009 Supplier Thermo Scientific Chemicals Supplier No. B2052009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29682-46-0 | C7H5Cl2NO2 | 206.02 g/mol

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 29682-46-0
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00024185
InChI Key WBNZUUIFTPNYRN-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
PubChem CID 97155
IUPAC Name 1,3-dichloro-2-methyl-4-nitrobenzene
SMILES CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O

Specifications

Melting Point 53°C to 55°C
Quantity 10 g
UN Number UN2811
Beilstein 2259855
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 99%
Chemical Name or Material 2,6-Dichloro-3-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 249-776-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only