missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-3-iodopyridine 98.0+%, TCI America™
$117.44 - $467.82
Chemical Identifiers
| CAS | 148493-37-2 |
|---|---|
| Molecular Formula | C5H2Cl2IN |
| Molecular Weight (g/mol) | 273.882 |
| MDL Number | MFCD03094686 |
| InChI Key | DPCQIHCGMIPSQV-UHFFFAOYSA-N |
| PubChem CID | 2736002 |
| IUPAC Name | 2,6-dichloro-3-iodopyridine |
| SMILES | C1=CC(=NC(=C1I)Cl)Cl |
Chemical Identifiers
| 148493-37-2 | |
| 273.882 | |
| DPCQIHCGMIPSQV-UHFFFAOYSA-N | |
| 2,6-dichloro-3-iodopyridine |
| C5H2Cl2IN | |
| MFCD03094686 | |
| 2736002 | |
| C1=CC(=NC(=C1I)Cl)Cl |
Specifications
| 148493-37-2 | |
| C5H2Cl2IN | |
| 1 g | |
| 2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine | |
| C1=CC(=NC(=C1I)Cl)Cl | |
| 273.882 | |
| 273.88 | |
| Crystalline Powder |
| 77°C | |
| MFCD03094686 | |
| 2811 | |
| DPCQIHCGMIPSQV-UHFFFAOYSA-N | |
| 2,6-dichloro-3-iodopyridine | |
| 2736002 | |
| ≥98.0% (GC) | |
| 2,6-Dichloro-3-iodopyridine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator