accessibility menu, dialog, popup

UserName

CAS RN 148493-37-2

IUPAC Name: 2,6-dichloro-3-iodopyridine
Synonyms: 2,6-dichloro-3-iodopyridine
Molecular Weight (g/mol): 273.88
Molecular Formula: C5H2Cl2IN
InChi Key: DPCQIHCGMIPSQV-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(I)C=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)