missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-iodopyridine 98.0+%, TCI America™

Catalog No. D50611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50611G 1 g
D50615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50611G Supplier TCI America Supplier No. D50611G

Chemical Identifiers

CAS 148493-37-2
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.882
MDL Number MFCD03094686
InChI Key DPCQIHCGMIPSQV-UHFFFAOYSA-N
PubChem CID 2736002
IUPAC Name 2,6-dichloro-3-iodopyridine
SMILES C1=CC(=NC(=C1I)Cl)Cl

Specifications

Melting Point 77°C
Quantity 1 g
UN Number 2811
Synonym 2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine
Formula Weight 273.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-3-iodopyridine
TSCA No
Recommended Storage Refrigerator