missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-iodopyridine, 98%, Thermo Scientific™

$122.16 - $122.16

Chemical Identifiers

CAS 148493-37-2
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.882
MDL Number MFCD03094686
InChI Key DPCQIHCGMIPSQV-UHFFFAOYSA-N
PubChem CID 2736002
IUPAC Name 2,6-dichloro-3-iodopyridine
SMILES C1=CC(=NC(=C1I)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6436606
View Documents
Thermo Scientific Chemicals
H6436606
5 g
Each for $122.16
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications

Chemical Identifiers

148493-37-2
273.882
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2,6-dichloro-3-iodopyridine
C5H2Cl2IN
MFCD03094686
2736002
C1=CC(=NC(=C1I)Cl)Cl

Specifications

148493-37-2
White to Brown
MFCD03094686
UN2811
Light sensitive
Slightly soluble in water.
C1=CC(=NC(=C1I)Cl)Cl
273.882
273.88
Powder
74°C to 79°C
C5H2Cl2IN
5 g
6135696
2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2,6-dichloro-3-iodopyridine
2736002
98%
2,6-Dichloro-3-iodopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'