missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloro-3'-fluoroacetophenone 97.0+%, TCI America™

Catalog No. D27505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27505G 5 g
D275025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27505G Supplier TCI America Supplier No. D27505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 290835-85-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD02093760
InChI Key VJBFZHHRVCPAPZ-UHFFFAOYSA-N
Synonym 2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
PubChem CID 2733982
IUPAC Name 1-(2,6-dichloro-3-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(Cl)C=CC(F)=C1Cl

Specifications

Color Yellow
Boiling Point 255°C
Quantity 5 g
Formula Weight 207.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,6′-Dichloro-3′-fluoroacetophenone