accessibility menu, dialog, popup

UserName

H3COClFCl

CAS RN 290835-85-7

IUPAC Name: 1-(2,6-dichloro-3-fluorophenyl)ethan-1-one
Synonyms: 1-(2,6-dichloro-3-fluorophenyl)ethanone
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H5Cl2FO
InChi Key: VJBFZHHRVCPAPZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)C=CC(F)=C1Cl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)