missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D03445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03445G 5 g
D03441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03445G Supplier TCI America Supplier No. D03445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 697-91-6
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.98
MDL Number MFCD00037159
InChI Key JCARTGJGWCGSSU-UHFFFAOYSA-N
Synonym 2,6-dichloro-1,4-benzoquinone, 2,6-dichloro-p-benzoquinone, 2,6-dichlorobenzoquinone, 2,6-dichloroquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dichloro, 2,6-dichloquinone, p-quinone, 2,6-dichloro, p-benzoquinone, 2,6-dichloro, 2,6-dichloro-2,5-cyclohexadiene-1,4-dione, p-quinone,6-dichloro
PubChem CID 12771
IUPAC Name 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C(=CC1=O)Cl)Cl

Specifications

Melting Point 122°C
Color Yellow
Quantity 5 g
Formula Weight 176.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-1,4-benzoquinone
RTECSNumber DK4000000
TSCA Yes