accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 697-91-6

IUPAC Name: 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
Synonyms: 2,6-dichlorobenzoquinone
Molecular Weight (g/mol): 176.98
Molecular Formula: C6H2Cl2O2
InChi Key: JCARTGJGWCGSSU-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)C=C(Cl)C1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)