missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromotoluene, 98+%

Catalog No. p-7081736
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1183203 1 g
AAA1183206 5 g
AAA1183214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1183203 Supplier Thermo Scientific Chemicals Supplier No. A1183203
Only null left

CAS: 69321-60-4 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69321-60-4
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013524
InChI Key OCSKCBIGEMSDIS-UHFFFAOYSA-N
Synonym 2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
PubChem CID 34681
IUPAC Name 1,3-dibromo-2-methylbenzene
SMILES CC1=C(Br)C=CC=C1Br

Specifications

Melting Point 2°C to 6°C
Density 1.812
Boiling Point 112°C to 113°C (7 mmHg)
Refractive Index 1.606
Quantity 1 g
Beilstein 3235502
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromotoluene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only