missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 449340050

 View more versions of this product

Catalog No. AC449340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 6-Dibromonaphthalene
98.5% min. (GC)
C10H6Br2
5g
Solubility in water: insoluble
BrC1=CC2=CC=C(Br)C=C2C=C1
285.97
285.97
13720-06-4
Glass bottle
MFCD01026462
naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
PJZDEYKZSZWFPX-UHFFFAOYSA-N
2,6-dibromonaphthalene
640591
99%

Chemical Identifiers

13720-06-4
285.97
PJZDEYKZSZWFPX-UHFFFAOYSA-N
640591
BrC1=CC2=CC=C(Br)C=C2C=C1
C10H6Br2
MFCD01026462
naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
2,6-dibromonaphthalene
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only