missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene, 99%

Catalog No. p-4812700
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449340010 1 g Glass bottle
AC449340050 5 g Glass bottle
AC449340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449340010 Supplier Thermo Scientific Chemicals Supplier No. 449340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13720-06-4 | C10H6Br2 | 285.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13720-06-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD01026462
InChI Key PJZDEYKZSZWFPX-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
PubChem CID 640591
IUPAC Name 2,6-dibromonaphthalene
SMILES BrC1=CC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 162°C to 167°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2, 6-Dibromonaphthalene

RUO – Research Use Only