missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoanthraquinone 95.0+%, TCI America™

Catalog No. D31821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31821G 1 g
D31825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31821G Supplier TCI America Supplier No. D31821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 633-70-5
Molecular Formula C14H6Br2O2
Molecular Weight (g/mol) 366.01
MDL Number MFCD08276344
InChI Key JUFYHUWBLXKCJM-UHFFFAOYSA-N
PubChem CID 10959484
IUPAC Name 2,6-dibromo-9,10-dihydroanthracene-9,10-dione
SMILES BrC1=CC2=C(C=C1)C(=O)C1=C(C=CC(Br)=C1)C2=O

Specifications

Melting Point 290°C
Quantity 1 g
Formula Weight 366.01
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromoanthraquinone