accessibility menu, dialog, popup

UserName

BrOOBr

CAS RN 633-70-5

IUPAC Name: 2,6-dibromo-9,10-dihydroanthracene-9,10-dione
Synonyms: 2,6-dibromoanthracene-9,10-dione
Molecular Weight (g/mol): 366.01
Molecular Formula: C14H6Br2O2
InChi Key: JUFYHUWBLXKCJM-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C(=O)C1=C(C=CC(Br)=C1)C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results