missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromoanisole 98.0+%, TCI America™
$141.67 - $423.14
Chemical Identifiers
| CAS | 38603-09-7 |
|---|---|
| Molecular Formula | C7H6Br2O |
| Molecular Weight (g/mol) | 265.93 |
| MDL Number | MFCD00093270 |
| InChI Key | BMZVDHQOAJUZJL-UHFFFAOYSA-N |
| Synonym | 1,3-Dibromo-2-methoxybenzene |
| PubChem CID | 181584 |
| IUPAC Name | 1,3-dibromo-2-methoxybenzene |
| SMILES | COC1=C(Br)C=CC=C1Br |
Chemical Identifiers
| 38603-09-7 | |
| 265.93 | |
| BMZVDHQOAJUZJL-UHFFFAOYSA-N | |
| 181584 | |
| COC1=C(Br)C=CC=C1Br |
| C7H6Br2O | |
| MFCD00093270 | |
| 1,3-Dibromo-2-methoxybenzene | |
| 1,3-dibromo-2-methoxybenzene |
Specifications
| 38603-09-7 | |
| Yellow | |
| C7H6Br2O | |
| 1 g | |
| BMZVDHQOAJUZJL-UHFFFAOYSA-N | |
| 1,3-dibromo-2-methoxybenzene | |
| 181584 | |
| ≥98.0% (GC) | |
| 2,6-Dibromoanisole |
| 13°C | |
| 145°C | |
| MFCD00093270 | |
| 1,3-Dibromo-2-methoxybenzene | |
| COC1=C(Br)C=CC=C1Br | |
| 265.93 | |
| 265.93 | |
| Liquid |
Safety and Handling
TSCA : No