missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromoanisole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 13°C |
| Color | Yellow |
| Boiling Point | 145°C |
| Quantity | 5 g |
| Formula Weight | 265.93 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,6-Dibromoanisole |
Safety and Handling
| TSCA | No |