missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-9,10-di(2-naphthyl)anthracene 98.0+%, TCI America™

Catalog No. d4646200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4646200MG 200 mg
D46461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4646200MG Supplier TCI America Supplier No. D4646200MG

Chemical Identifiers

CAS 561064-15-1
Molecular Formula C34H20Br2
Molecular Weight (g/mol) 588.34
MDL Number MFCD16660916
InChI Key WQURYVBAXSWGDX-UHFFFAOYSA-N
PubChem CID 22497802
IUPAC Name 2,6-dibromo-9,10-bis(naphthalen-2-yl)anthracene
SMILES BrC1=CC2=C(C3=CC=C4C=CC=CC4=C3)C3=CC=C(Br)C=C3C(C3=CC4=CC=CC=C4C=C3)=C2C=C1

Specifications

Quantity 200 mg
Formula Weight 588.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-9,10-di(2-naphthyl)anthracene