accessibility menu, dialog, popup

UserName

CAS RN 561064-15-1

IUPAC Name: 2,6-dibromo-9,10-bis(naphthalen-2-yl)anthracene
Synonyms: 2,6-dibromo-9,10-bis(naphthalen-2-yl)anthracene
Molecular Weight (g/mol): 588.34
Molecular Formula: C34H20Br2
InChi Key: WQURYVBAXSWGDX-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C3=CC=C4C=CC=CC4=C3)C3=CC=C(Br)C=C3C(C3=CC4=CC=CC=C4C=C3)=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results