missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%

Catalog No. p-7045425
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1065803 1 g
AAL1065806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1065803 Supplier Thermo Scientific Chemicals Supplier No. L1065803

CAS: 72678-19-4 | C7H4Br2F3N | 318.919 g/mol

Used in enantio-and diastereoselective addition reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in enantio-and diastereoselective addition reation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 72678-19-4
Molecular Formula C7H4Br2F3N
Molecular Weight (g/mol) 318.919
MDL Number MFCD00068181
InChI Key DRSMEHXBOXHXDX-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl-phenylamine
PubChem CID 144565
IUPAC Name 2,6-dibromo-4-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F

Specifications

Melting Point 36°C to 40°C
Boiling Point 64°C to 65°C (0.1 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 6314196
Solubility Information Insoluble in water.
Formula Weight 318.93
Percent Purity 97%
Chemical Name or Material 2,6-Dibromo-4-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only