missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-nitrophenol, 98%

Catalog No. aaa1291622
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1291622 100 g
AAA1291614 25 g
AAA1291630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1291622 Supplier Thermo Scientific Chemicals Supplier No. A1291622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-28-5 | C6H3Br2NO3 | 296.902 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-28-5
Molecular Formula C6H3Br2NO3
Molecular Weight (g/mol) 296.902
MDL Number MFCD00007334
InChI Key WBHYZUAQCSHXCT-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-nitrophenol, phenol, 2,6-dibromo-4-nitro, 4-nitro-2,6-dibromophenol, 2,6-dibromo-4-nitro-phenol, pubchem3863, acmc-209sc6, wln: wnr dq ce ee, phenol,6-dibromo-4-nitro, 4-06-00-01366 beilstein handbook reference, 2,6-dibromo-4-nitro-phenolate
PubChem CID 7429
SMILES C1=C(C=C(C(=C1Br)O)Br)[N+](=O)[O-]

Specifications

Melting Point ∼145°C (decomposition)
Quantity 100 g
Beilstein 1245050
Formula Weight 296.9
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-nitrophenol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 202-744-3
RTECSNumber SK8025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only