missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-nitroaniline 98.0+%, TCI America™
$98.18 - $854.77
Chemical Identifiers
| CAS | 827-94-1 |
|---|---|
| Molecular Formula | C6H4Br2N2O2 |
| Molecular Weight (g/mol) | 295.92 |
| MDL Number | MFCD00007639 |
| InChI Key | YMZIFDLWYUSZCC-UHFFFAOYSA-N |
| Synonym | benzenamine, 2,6-dibromo-4-nitro, aniline, 2,6-dibromo-4-nitro, unii-gwi915pon9, gwi915pon9, 2,6-dibromo-4-nitro-aniline, 2,6-dibromo-4-nitrophenylamine, 2,6-dibromo-4-nitro-phenylamine, pubchem2330, dsstox_cid_31146, dsstox_gsid_52573 |
| PubChem CID | 13231 |
| IUPAC Name | 2,6-dibromo-4-nitroaniline |
| SMILES | NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O |
Chemical Identifiers
| 827-94-1 | |
| 295.92 | |
| YMZIFDLWYUSZCC-UHFFFAOYSA-N | |
| 13231 | |
| NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O |
| C6H4Br2N2O2 | |
| MFCD00007639 | |
| benzenamine, 2,6-dibromo-4-nitro, aniline, 2,6-dibromo-4-nitro, unii-gwi915pon9, gwi915pon9, 2,6-dibromo-4-nitro-aniline, 2,6-dibromo-4-nitrophenylamine, 2,6-dibromo-4-nitro-phenylamine, pubchem2330, dsstox_cid_31146, dsstox_gsid_52573 | |
| 2,6-dibromo-4-nitroaniline |
Specifications
| 827-94-1 | |
| Yellow | |
| MFCD00007639 | |
| benzenamine, 2,6-dibromo-4-nitro, aniline, 2,6-dibromo-4-nitro, unii-gwi915pon9, gwi915pon9, 2,6-dibromo-4-nitro-aniline, 2,6-dibromo-4-nitrophenylamine, 2,6-dibromo-4-nitro-phenylamine, pubchem2330, dsstox_cid_31146, dsstox_gsid_52573 | |
| NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O | |
| 295.92 | |
| 295.92 | |
| Crystalline Powder |
| 207°C | |
| C6H4Br2N2O2 | |
| 25 g | |
| YMZIFDLWYUSZCC-UHFFFAOYSA-N | |
| 2,6-dibromo-4-nitroaniline | |
| 13231 | |
| ≥98.0% (GC) | |
| 2,6-Dibromo-4-nitroaniline |
Safety and Handling
EINECSNumber : (3)-0424
RTECSNumber : BX2599000
TSCA : Yes